siRNA Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029436.3 | NP_083712.4 | 7 | 1668 | ||
| GenBank mRNA | AK015239.1 | - | 7 | 1654 | ||
| AK044892.1 | - | 7 | 1640 | |||
| AK046446.1 | - | 5 | 746 | |||
| AK163616.1 | - | 5 | 806 | |||
| BC021407.1 | - | 5 | 1222 | |||

Klhl24
Genomic Map