siRNA Details
| Gene Symbol: Tasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159640.2 | NP_001153112.1 | 4 | 349 | ||
| NM_001159641.2 | NP_001153113.1 | 4 | 349 | |||
| NM_001289611.1 | NP_001276540.1 | 4 | 349 | |||
| NM_175225.5 | NP_780434.1 | 4 | 616 | |||
| GenBank mRNA | AK015624.1 | - | 4 | 349 | ||
| AK036512.1 | - | 4 | 572 | |||
| AK050603.1 | - | 1 | 73 | |||
| AK146320.1 | - | 4 | 332 | |||
| AK160983.1 | - | 4 | 577 | |||
| BB665115.1 | - | 4 | 325 | |||
| BC024597.1 | - | 4 | 336 | 4036044 | ||
| BY055918.1 | - | 4 | 329 | |||

Tasp1
Genomic Map