siRNA Details
| Gene Symbol: Senp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029457.3 | NP_083733.1 | 13 | 1474 | ||
| GenBank mRNA | AF368904.1 | - | 13 | 1520 | ||
| AK015987.1 | - | 13 | 1453 | |||
| AK031030.1 | - | 13 | 1459 | |||
| AY188288.1 | - | 12 | 1246 | |||
| BC031652.1 | - | 13 | 1449 | 3483046 | ||
| BC040805.1 | - | 5 | 1418 | |||
| BC042514.1 | - | 5 | 1418 | |||

Senp2
Genomic Map