siRNA Details
| Gene Symbol: 4930578C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175228.3 | NP_780437.1 | 2 | 354 | ||
| GenBank mRNA | AK016298.1 | - | 2 | 359 | ||
| AK085770.1 | - | 2 | 355 | |||
| AK163928.1 | - | 4 | 697 | |||
| BC118515.1 | - | 2 | 355 | 40105826 | ||

4930578C19Rik
Genomic Map