siRNA Details
Gene Symbol: Samd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029489.3 | NP_083765.2 | 6 | 1074 | ||
GenBank mRNA | AK044227.1 | - | 5 | 950 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Samd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029489.3 | NP_083765.2 | 6 | 1074 | ||
GenBank mRNA | AK044227.1 | - | 5 | 950 |