siRNA Details
| Gene Symbol: Dock11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009947.3 | NP_001009947.2 | 25 | 2834 | ||
| GenBank mRNA | AB116935.2 | - | 25 | 2834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock11| Gene Symbol: Dock11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009947.3 | NP_001009947.2 | 25 | 2834 | ||
| GenBank mRNA | AB116935.2 | - | 25 | 2834 | ||