siRNA Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081408.1 | NP_001074877.1 | 6, 7 | 1132 | ||
| GenBank mRNA | AK017155.1 | - | 6, 7 | 1029 | ||
| BC025916.1 | - | 3, 4 | 349 | |||
| BC037459.1 | - | 3, 4 | 305 | |||
| BC115693.2 | - | 6, 7 | 1029 | 40104399 | ||

Agmat
Genomic Map