siRNA Details
| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029498.3 | NP_083774.2 | 5 | 1450 | ||
| GenBank mRNA | AK135499.1 | - | 6 | 1563 | ||
| BC043450.1 | - | 2 | 215 | |||
| BC084596.1 | - | 1 | 57 | |||
| BC141403.1 | - | 5 | 1426 | 9056016 | ||

Zmym2
Genomic Map