siRNA Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168386.2 | NP_001161858.1 | 2, 3 | 663 | ||
| NM_183115.5 | NP_898938.4 | 2, 3 | 663 | |||
| GenBank mRNA | AK017973.1 | - | 2, 3 | 426 | ||
| BC085249.1 | - | 2, 3 | 381 | 30726150 | ||
| BU513278.1 | - | 3, 4 | 771 | |||

Ccdc125
Genomic Map