siRNA Details
| Gene Symbol: Ncapg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133762.3 | NP_598523.3 | 13 | 1515 | ||
| GenBank mRNA | AY455829.1 | - | 7 | 626 | ||
| BC008539.1 | - | 2 | 196 | |||
| BC076631.1 | - | 12 | 1325 | 30637859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncapg2| Gene Symbol: Ncapg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133762.3 | NP_598523.3 | 13 | 1515 | ||
| GenBank mRNA | AY455829.1 | - | 7 | 626 | ||
| BC008539.1 | - | 2 | 196 | |||
| BC076631.1 | - | 12 | 1325 | 30637859 | ||