siRNA Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178394.4 | NP_848481.2 | 4 | 1063 | ||
| GenBank mRNA | AK017977.1 | - | 4 | 1018 | ||
| AK077298.1 | - | 4 | 1005 | |||
| BC138646.1 | - | 3 | 915 | 8861666 | ||
| BC138649.1 | - | 3 | 915 | 8861669 | ||

Jakmip1
Genomic Map