siRNA Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178394.4 | NP_848481.2 | 9, 10 | 1676 | ||
| GenBank mRNA | AK077298.1 | - | 9, 10 | 1618 | ||
| BC138646.1 | - | 8, 9 | 1528 | 8861666 | ||
| BC138649.1 | - | 8, 9 | 1528 | 8861669 | ||

Jakmip1
Genomic Map