siRNA Details
| Gene Symbol: Ccdc138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162956.1 | NP_001156428.1 | 8 | 913 | ||
| GenBank mRNA | BC119028.1 | - | 8 | 902 | 8733777 | |
| BC119030.1 | - | 8 | 902 | 8733779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc138| Gene Symbol: Ccdc138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162956.1 | NP_001156428.1 | 8 | 913 | ||
| GenBank mRNA | BC119028.1 | - | 8 | 902 | 8733777 | |
| BC119030.1 | - | 8 | 902 | 8733779 | ||