siRNA Details
Gene Symbol: Fam131b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113327.2 | NP_001106798.1 | 5, 6 | 745 | ||
NM_001286584.1 | NP_001273513.1 | 6, 7 | 721 | |||
NM_029528.5 | NP_083804.3 | 6, 7 | 781 | |||
GenBank mRNA | AK018206.1 | - | 6, 7 | 720 | ||
AK018325.1 | - | 6, 7 | 727 | |||
AK047763.1 | - | 6, 7 | 707 | |||
AK158869.1 | - | 6, 7 | 674 | |||
AK163654.1 | - | 5, 6 | 666 | |||
BC115595.1 | - | 5, 6 | 511 | |||
BC115596.1 | - | 5, 6 | 511 |
