siRNA Details
| Gene Symbol: Abca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147218.2 | NP_671751.2 | 39 | 5020 | ||
| GenBank mRNA | AF498361.1 | - | 39 | 4972 | ||
| BC132417.1 | - | 39 | 4857 | 40130694 | ||
| BC132419.1 | - | 39 | 4857 | 40130696 | ||

Abca6
Genomic Map