siRNA Details
| Gene Symbol: Abca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166556.1 | NP_001160028.1 | 7 | 1092 | ||
| NM_001166557.1 | NP_001160029.1 | 7 | 1092 | |||
| NM_147218.2 | NP_671751.2 | 7 | 1092 | |||
| GenBank mRNA | AF498361.1 | - | 7 | 1044 | ||
| AK018242.1 | - | 7 | 1077 | |||
| AK040652.1 | - | 7 | 1092 | |||
| AK042934.1 | - | 7 | 1064 | |||
| AK053384.1 | - | 7 | 1019 | |||
| BC132417.1 | - | 7 | 929 | 40130694 | ||
| BC132419.1 | - | 7 | 929 | 40130696 | ||

Abca6
Genomic Map