siRNA Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175236.4 | NP_780445.1 | 12 | 1188 | ||
| GenBank mRNA | AK018241.1 | - | 12 | 1159 | ||
| AK038853.1 | - | 12 | 1155 | |||
| AK046780.1 | - | 12 | 1114 | |||
| AK050178.1 | - | 11 | 1112 | |||
| AK084104.1 | - | 1117 | ||||
| BC026584.1 | - | 12 | 1146 | 4972323 | ||

Adhfe1
Genomic Map