siRNA Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175236.4 | NP_780445.1 | 5 | 312 | ||
| GenBank mRNA | AK018241.1 | - | 5 | 286 | ||
| AK038853.1 | - | 5 | 279 | |||
| AK046780.1 | - | 5 | 238 | |||
| AK050178.1 | - | 4 | 236 | |||
| AK084104.1 | - | 239 | ||||
| BC026584.1 | - | 5 | 270 | 4972323 | ||
| CA467848.1 | - | 5 | 351 | |||

Adhfe1
Genomic Map