siRNA Details
| Gene Symbol: Med13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172424.4 | NP_766012.3 | 20 | 4503 | ||
| GenBank mRNA | AK122419.1 | - | 11 | 2791 | ||
| AK154895.1 | - | 20 | 4483 | |||
| AY338464.1 | - | 20 | 4480 | |||
| BC052320.1 | - | 4 | 1190 | |||
| BC072653.1 | - | 13 | 3426 | |||

Med13l
Genomic Map