siRNA Details
| Gene Symbol: Ttc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029553.3 | NP_083829.1 | 13 | 1430 | ||
| NM_198311.1 | NP_938053.1 | 14 | 1422 | |||
| GenBank mRNA | AK002597.1 | - | 13 | 1416 | ||
| AK032748.1 | - | 13 | 1392 | |||
| AK081697.1 | - | 13 | 1397 | |||
| BC017523.1 | - | 14 | 1422 | 4527657 | ||

Ttc8
Genomic Map