siRNA Details
| Gene Symbol: Tsen54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029557.1 | NP_083833.1 | 9 | 1286 | ||
| GenBank mRNA | AK002758.1 | - | 9 | 1271 | ||
| AK089005.1 | - | 9 | 1253 | |||
| BC051543.1 | - | 2 | 267 | |||
| BC117762.1 | - | 3 | 341 | |||
| BC126953.1 | - | 8 | 1410 | |||

Tsen54
Genomic Map