siRNA Details
| Gene Symbol: Atp11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029570.3 | NP_083846.2 | 7 | 859 | ||
| GenBank mRNA | AK129248.1 | - | 7 | 851 | ||
| AK154156.1 | - | 7 | 863 | |||
| AK155372.1 | - | 7 | 863 | |||
| BC076603.1 | - | 7 | 729 | 30629594 | ||

Atp11b
Genomic Map