siRNA Details
| Gene Symbol: Rab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029576.3 | NP_083852.1 | 6 | 515 | ||
| GenBank mRNA | AB232584.1 | - | 5 | 457 | ||
| AK003609.1 | - | 6 | 516 | |||
| AK042020.1 | - | 6 | 517 | |||
| AK156726.1 | - | 6 | 538 | |||
| AK200798.1 | - | 2 | 164 | |||
| BC016408.1 | - | 6 | 510 | 4242206 | ||

Rab1b
Genomic Map