siRNA Details
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029576.3 | NP_083852.1 | 5 | 391 | ||
GenBank mRNA | AB232584.1 | - | 4 | 333 | ||
AK003609.1 | - | 5 | 392 | |||
AK042020.1 | - | 5 | 393 | |||
AK156726.1 | - | 5 | 414 | |||
AK200798.1 | - | 1 | 40 | |||
BC016408.1 | - | 5 | 386 | 4242206 |
