siRNA Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029578.3 | NP_083854.3 | 2 | 154 | ||
| GenBank mRNA | AK011439.1 | - | 2 | 133 | ||
| AK011555.1 | - | 2 | 145 | |||
| AK154341.1 | - | 2 | 131 | |||
| AK165685.1 | - | 2 | 128 | |||
| AK165955.1 | - | 2 | 138 | |||
| BC021419.1 | - | 2 | 130 | 5012877 | ||
| BY710811.1 | - | 2 | 142 | |||
| CB953719.1 | - | 2 | 157 | |||

Tgds
Genomic Map