siRNA Details
| Gene Symbol: Ropn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030744.2 | NP_109669.1 | 4 | 458 | ||
| GenBank mRNA | AF178531.1 | - | 3 | 381 | ||
| AK160142.1 | - | 2 | 200 | |||
| BC049544.1 | - | 4 | 458 | 6740976 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ropn1| Gene Symbol: Ropn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030744.2 | NP_109669.1 | 4 | 458 | ||
| GenBank mRNA | AF178531.1 | - | 3 | 381 | ||
| AK160142.1 | - | 2 | 200 | |||
| BC049544.1 | - | 4 | 458 | 6740976 | ||