siRNA Details
| Gene Symbol: Abcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029600.3 | NP_083876.3 | 15, 16 | 1993 | ||
| GenBank mRNA | AF534127.1 | - | 15, 16 | 1993 | ||
| AF534128.1 | - | 15, 16 | 1993 | |||
| AF534129.1 | - | 15, 16 | 1993 | |||
| AK149551.1 | - | 15, 16 | 1993 | |||
| AK155664.1 | - | 15, 16 | 1968 | |||
| AK156615.1 | - | 15, 16 | 2223 | |||
| AK172420.1 | - | 15, 16 | 2223 | |||
| AY795569.1 | - | 15, 16 | 1993 | |||
| AY841885.1 | - | 15, 16 | 1974 | |||
| BC043728.1 | - | 4, 5 | 1081 | |||
| BC046560.1 | - | 12, 13 | 1497 | |||
| BC048825.1 | - | 15, 16 | 1925 | |||
| BC150788.1 | - | 15, 16 | 1984 | 9087699 | ||

Abcc3
Genomic Map