siRNA Details
Gene Symbol: 2310001H17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009064.1 | - | 1 | 117 | ||
AK030376.1 | - | 83 | ||||
AV065335.2 | - | 1 | 206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310001H17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009064.1 | - | 1 | 117 | ||
AK030376.1 | - | 83 | ||||
AV065335.2 | - | 1 | 206 |