siRNA Details
| Gene Symbol: Daam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008231.2 | NP_001008232.2 | 9, 10 | 1231 | ||
| GenBank mRNA | AK122266.1 | - | 9, 10 | 1231 | ||
| AY426536.1 | - | 9, 10 | 1260 | |||
| BC050043.1 | - | 9, 10 | 1231 | |||

Daam2
Genomic Map