siRNA Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029624.4 | NP_083900.3 | 7 | 1055 | ||
| GenBank mRNA | AK010312.1 | - | 7 | 1026 | ||
| AK041659.1 | - | 5 | 722 | |||
| AK154622.1 | - | 7 | 1052 | |||
| AK169983.1 | - | 7 | 1053 | |||
| AK172374.1 | - | 7 | 928 | |||
| AK177501.1 | - | 2 | 187 | |||
| AK183502.1 | - | 2 | 187 | |||
| BC020104.1 | - | 7 | 1062 | 3963699 | ||

Lmf1
Genomic Map