siRNA Details
| Gene Symbol: Trappc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164641.1 | NP_001158113.1 | 6 | 1187 | ||
| NM_001164642.1 | NP_001158114.1 | 5 | 1160 | |||
| NM_001164643.1 | NP_001158115.1 | 5 | 1160 | |||
| NM_029640.2 | NP_083916.1 | 5 | 415 | |||
| NM_180662.2 | NP_850993.2 | 5 | 1160 | |||
| GenBank mRNA | AK028504.1 | - | 5 | 1002 | ||
| AK029106.1 | - | 6 | 1029 | |||
| AK031788.1 | - | 5 | 1008 | |||
| AK138682.1 | - | 5 | 1007 | |||
| AK156739.1 | - | 5 | 985 | |||
| AK214984.1 | - | 2 | 131 | |||
| AY630620.1 | - | 5 | 413 | |||
| BC034590.1 | - | 5 | 413 | 5364921 | ||
| BC070463.1 | - | 5 | 1160 | 30613261 | ||

Trappc9
Genomic Map