siRNA Details
| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029648.6 | NP_083924.2 | 2 | 564 | ||
| GenBank mRNA | AK011404.2 | - | 2 | 543 | ||
| AK075914.1 | - | 2 | 565 | |||
| AK162079.1 | - | 2 | 516 | |||
| BC048505.1 | - | 3 | 191 | |||
| BC061459.1 | - | 363 | 6829614 | |||

Fam204a
Genomic Map