siRNA Details
| Gene Symbol: Atg2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029654.4 | NP_083930.5 | 38 | 5940 | ||
| GenBank mRNA | AK220468.1 | - | 2 | 3975 | ||
| BC024533.1 | - | 6 | 562 | |||
| BC046427.1 | - | 17 | 2051 | |||
| BC062182.1 | - | 38 | 5910 | 6505402 | ||

Atg2b
Genomic Map