siRNA Details
| Gene Symbol: Atg2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029654.4 | NP_083930.5 | 41 | 6390 | ||
| GenBank mRNA | AK010577.1 | - | 4 | 554 | ||
| AK220468.1 | - | 4 | 5017 | |||
| BC024533.1 | - | 9 | 1012 | |||
| BC046427.1 | - | 20 | 2501 | |||
| BC062182.1 | - | 41 | 6360 | 6505402 | ||
| CA751117.1 | - | 4 | 435 | |||

Atg2b
Genomic Map