siRNA Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023831.3 | NP_076320.2 | 10 | 974 | ||
| GenBank mRNA | AK005360.1 | - | 10 | 982 | ||
| AK032880.1 | - | 10 | 975 | |||
| BC010326.1 | - | 9 | 773 | 3599905 | ||
| BC080764.1 | - | 10 | 916 | 6488976 | ||

Ift46
Genomic Map