siRNA Details
| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289554.1 | NP_001276483.1 | 7 | 773 | ||
| NM_001289555.1 | NP_001276484.1 | 7 | 770 | |||
| NM_001289556.1 | NP_001276485.1 | 6 | 614 | |||
| NM_001289557.1 | NP_001276486.1 | 5 | 465 | |||
| NM_029659.4 | NP_083935.3 | 7 | 780 | |||
| GenBank mRNA | AK005599.1 | - | 7 | 771 | ||
| AK005858.1 | - | 6 | 521 | |||
| AK161310.1 | - | 7 | 781 | |||
| BC045148.1 | - | 7 | 710 | 6307739 | ||
| BF471066.1 | - | 5 | 374 | |||

Styxl1
Genomic Map