siRNA Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152823.4 | NP_690036.4 | 9 | 1670 | ||
| GenBank mRNA | AK010945.1 | - | 2 | 332 | ||
| AK089034.1 | - | 5 | 539 | |||
| AK144925.1 | - | 9 | 1672 | |||
| AY510108.1 | - | 8 | 1537 | |||
| BC022701.1 | - | 5 | 624 | |||
| BC095959.1 | - | 9 | 1648 | 5349225 | ||

Unc5cl
Genomic Map