siRNA Details
| Gene Symbol: Hectd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175244.3 | NP_780453.1 | 20, 21 | 2553 | ||
| GenBank mRNA | AK006883.1 | - | 1, 2 | 23 | ||
| AK051385.1 | - | 20, 21 | 2566 | |||
| AK139305.1 | - | 20, 21 | 2581 | |||
| AK154378.1 | - | 20, 21 | 2585 | |||
| AK155290.1 | - | 20, 21 | 2566 | |||
| AK155676.1 | - | 20, 21 | 2566 | |||
| AK163323.1 | - | 20, 21 | 2588 | |||
| AK165482.1 | - | 20, 21 | 2585 | |||
| AK170930.1 | - | 20, 21 | 2566 | |||
| BC024415.1 | - | 6, 7 | 621 | |||
| BC070411.1 | - | 20, 21 | 2553 | 5706209 | ||
| BC082276.1 | - | 10, 11 | 1007 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764069.

Hectd3
Genomic Map