siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143755.1 | NP_001137227.1 | 7 | 1115 | ||
| NM_027164.1 | NP_081440.1 | 6 | 1160 | |||
| GenBank mRNA | AK006931.1 | - | 7 | 1119 | ||
| BC050816.1 | - | 6 | 1160 | 6774506 | ||
| CO040070.1 | - | 3 | 589 | |||

Lrrc27
Genomic Map