siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143755.1 | NP_001137227.1 | 5 | 677 | ||
| NM_027164.1 | NP_081440.1 | 4 | 722 | |||
| GenBank mRNA | AK006931.1 | - | 5 | 682 | ||
| BC050816.1 | - | 4 | 722 | 6774506 | ||

Lrrc27
Genomic Map