siRNA Details
Gene Symbol: Msi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054043.3 | NP_473384.1 | 10 | 807 | ||
GenBank mRNA | AK049637.1 | - | 10 | 807 | ||
AK049688.1 | - | 10 | 807 | |||
AK051269.1 | - | 10 | 657 | |||
AK081959.1 | - | 1 | 1776 | |||
AK136156.1 | - | 6 | 615 | |||
AK153928.1 | - | 10 | 680 | |||
AK169846.1 | - | 10 | 680 | |||
BC111809.1 | - | 11 | 740 | 40045350 |
