siRNA Details
| Gene Symbol: Snx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082484.2 | NP_001075953.1 | 3 | 876 | ||
| NM_029721.2 | NP_083997.1 | 3 | 876 | |||
| GenBank mRNA | AK122296.1 | - | 3 | 688 | ||
| AK147452.1 | - | 3 | 863 | |||
| BC053495.1 | - | 3 | 688 | |||

Snx27
Genomic Map