siRNA Details
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029742.2 | NP_084018.1 | 10 | 1375 | ||
GenBank mRNA | AK010783.1 | - | 10 | 1240 | ||
AK039732.1 | - | 9 | 1277 | |||
AK050289.1 | - | 8 | 996 | |||
AK053834.1 | - | 9 | 1247 | |||
AK085116.1 | - | 9 | 1326 | |||
AK085130.1 | - | 10 | 1333 | |||
AK172923.1 | - | 9 | 1288 | |||
BC057064.1 | - | 5 | 2828 | |||
BC060132.1 | - | 10 | 1375 | 6409751 |
