siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029742.2 | NP_084018.1 | 5 | 727 | ||
| GenBank mRNA | AK010783.1 | - | 5 | 592 | ||
| AK039732.1 | - | 4 | 629 | |||
| AK050289.1 | - | 3 | 348 | |||
| AK053834.1 | - | 4 | 599 | |||
| AK085116.1 | - | 4 | 678 | |||
| AK085130.1 | - | 5 | 685 | |||
| AK172923.1 | - | 4 | 640 | |||
| BC057064.1 | - | 1 | 678 | |||
| BC060132.1 | - | 5 | 727 | 6409751 | ||

Klhdc10
Genomic Map