siRNA Details
| Gene Symbol: Tbc1d9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290759.1 | NP_001277688.1 | 4 | 549 | ||
| NM_001290760.1 | NP_001277689.1 | 4 | 549 | |||
| NM_029745.2 | NP_084021.2 | 4 | 549 | |||
| GenBank mRNA | AK035457.1 | - | 4 | 514 | ||
| AK049999.1 | - | 4 | 509 | |||
| AK147964.1 | - | 4 | 514 | |||
| AK166472.1 | - | 4 | 532 | |||
| AK186143.1 | - | 3 | 359 | |||
| BC062928.1 | - | 4 | 536 | 6854669 | ||
| BC065080.1 | - | 4 | 528 | 6826791 | ||
| BC065130.1 | - | 4 | 494 | |||

Tbc1d9b
Genomic Map