siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 7, 8 | 1077 | ||
| GenBank mRNA | AK012392.1 | - | 7, 8 | 865 | ||
| AK017691.1 | - | 7, 8 | 860 | |||
| AK051683.1 | - | 7, 8 | 880 | |||
| AK132316.1 | - | 7, 8 | 873 | |||
| AK133622.1 | - | 7, 8 | 821 | |||
| AK136921.1 | - | 7, 8 | 900 | |||
| BC016244.1 | - | 7, 8 | 857 | 4507575 | ||
| BC025138.1 | - | 6, 7 | 733 | 4483636 | ||

Sdccag8
Genomic Map