siRNA Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB095735.1 | - | 13 | 1303 | ||
| AB095736.1 | - | 13 | 1303 | |||
| AK054412.1 | - | 13 | 1293 | |||
| AK133177.1 | - | 13 | 1302 | |||
| AK154594.1 | - | 14 | 1343 | |||
| AK161401.1 | - | 10 | 904 | |||
| AK164249.1 | - | 13 | 1303 | |||
| AK166351.1 | - | 13 | 1311 | |||
| AK169601.1 | - | 13 | 1302 | |||
| BC060208.1 | - | 13 | 1262 | |||

Dtl
Genomic Map