siRNA Details
| Gene Symbol: Catsper3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252487.1 | NP_001239416.1 | 5, 6 | 897 | ||
| NM_001252488.1 | NP_001239417.1 | 5, 6 | 744 | |||
| NM_029772.4 | NP_084048.1 | 5, 6 | 858 | |||
| GenBank mRNA | AK014942.1 | - | 5, 6 | 859 | ||
| AY263400.1 | - | 5, 6 | 872 | |||
| BC089518.1 | - | 997 | 6774298 | |||
| HM179540.1 | - | 5, 6 | 814 | |||
| HM179541.1 | - | 5, 6 | 661 | |||

Catsper3
Genomic Map