siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163328.1 | NP_001156800.1 | 2 | 474 | ||
| NM_178631.4 | NP_848746.1 | 3 | 567 | |||
| GenBank mRNA | AK032086.1 | - | 2 | 407 | ||
| AK079659.1 | - | 2 | 1296 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK003003.1 | - | 282 | Chr. - - [-] on Build | ||

Ralyl
Genomic Map