siRNA Details
Gene Symbol: Ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163328.1 | NP_001156800.1 | 8, 9 | 1175 | ||
NM_001163329.1 | NP_001156801.1 | 7, 8 | 768 | |||
NM_178631.4 | NP_848746.1 | 9, 10 | 1268 | |||
GenBank mRNA | AK032086.1 | - | 8, 9 | 1108 | ||
AK038465.1 | - | 7, 8 | 746 | |||
AK079659.1 | - | 8, 9 | 1997 | |||
BC052358.1 | - | 7, 8 | 729 | 6534677 |
